KEY087886591 Key Organics / BIONET

ethyl (2E)-2-[(carbamoylamino)imino]-3-[(4-chlorophenyl)sulfanyl]propanoate

  • MDL number MFCD08056619
  • Cas Number : 886361-57-5
  • Molecular Weight : 315.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153 - 156
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70712

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3G-332S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart