KEY087886594 Key Organics / BIONET

4-chloro-2-{[(4-chlorophenyl)methyl]sulfanyl}-6-(methoxymethyl)pyrimidine

  • MDL number MFCD08056621
  • Cas Number : 886361-61-1
  • Molecular Weight : 315.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80 - 82
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71955

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3M-570S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart