KEY087886597 Key Organics / BIONET

(2,5-dichloropyridin-3-yl)methanol

  • MDL number MFCD06659487
  • Cas Number : 558465-93-3
  • Molecular Weight : 178.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 64 - 66
Iupac Name (2,5-dichloro-3-pyridinyl)methanol
Inchi Code 1S/C6H5Cl2NO/c7-5-1-4(3-10)6(8)9-2-5/h1-2,10H,3H2
Inchi Key OMVBAPGBNPDUNI-UHFFFAOYSA-N

Description

Melting Point 64 - 66
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73658

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3X-0704_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart