KEY087886615 Key Organics / BIONET

ethyl 5-phenylthiophene-2-carboxylate

  • MDL number MFCD01860759
  • Cas Number : 19282-39-4
  • Molecular Weight : 232.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 31 - 33
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77512

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4Z-0717_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart