KEY087886617 Key Organics / BIONET

2-[2-methyl-5-(propan-2-yl)phenoxy]benzaldehyde

  • MDL number MFCD07698594
  • Cas Number : 886361-81-5
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 142 @ 0.2 mBAR
Iupac Name 2-(5-isopropyl-2-methylphenoxy)benzaldehyde
Inchi Code 1S/C17H18O2/c1-12(2)14-9-8-13(3)17(10-14)19-16-7-5-4-6-15(16)11-18/h4-12H,1-3H3
Inchi Key SJHOEARKFOIYOT-UHFFFAOYSA-N

Description

Melting Point 142 @ 0.2 mBAR
Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77517

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4Z-0801_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart