KEY087886620 Key Organics / BIONET

3-amino-3-(3-fluoro-4-methylphenyl)propanoic acid

  • MDL number MFCD03002496
  • Cas Number : 682804-13-3
  • Molecular Weight : 197.209

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 233 - 235DEC
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77529

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4Z-0829_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart