KEY087886630 Key Organics / BIONET

2-(5-methyl-1H-pyrazol-3-yl)-1,3-thiazole

  • MDL number MFCD08056642
  • Cas Number : 1187559-72-3
  • Molecular Weight : 165.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126 - 128
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81142

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5Z-0813_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart