KEY087886633 Key Organics / BIONET

3-hydrazinyl-5-(trifluoromethyl)-4H-1,2,4-triazol-4-amine hydrochloride

  • MDL number MFCD08056644
  • Cas Number : 1172756-64-7
  • Molecular Weight : 218.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165 - 168
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82391

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6K-064_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart