KEY087886639 Key Organics / BIONET

ethyl (2E)-3-[4-(propan-2-yl)phenyl]prop-2-enoate

  • MDL number MFCD08056649
  • Cas Number : 32580-69-1
  • Molecular Weight : 218.296

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84504

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6Z-0260_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart