KEY087886641 Key Organics / BIONET

3-(2H-1,3-benzodioxol-5-yl)-3-{1,4-dioxa-8-azaspiro[4.5]decan-8-yl}propanoic acid

  • MDL number MFCD08056651
  • Cas Number : 886361-92-8
  • Molecular Weight : 335.356

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191 - 193
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84506

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6Z-0276_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart