KEY087886648 Key Organics / BIONET

[2-(4-chlorophenoxy)-5-nitrophenyl]methanol

  • MDL number MFCD08056656
  • Cas Number : 886361-95-1
  • Molecular Weight : 279.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118 - 120
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84523

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6Z-0814_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart