KEY087886649 Key Organics / BIONET

5-methoxy-1-(pyrimidin-2-yl)-1H-indole-3-carbaldehyde

  • MDL number MFCD08056657
  • Cas Number : 886361-96-2
  • Molecular Weight : 253.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 150 - 152
Iupac Name 5-methoxy-1-(2-pyrimidinyl)-1H-indole-3-carbaldehyde
Inchi Code 1S/C14H11N3O2/c1-19-11-3-4-13-12(7-11)10(9-18)8-17(13)14-15-5-2-6-16-14/h2-9H,1H3
Inchi Key DYXNNMOTBJZTPI-UHFFFAOYSA-N

Description

Melting Point 150 - 152
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84525

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6Z-0825_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart