KEY087886653 Key Organics / BIONET

(2E)-1-(4-bromophenyl)-3-(dimethylamino)but-2-en-1-one

  • MDL number MFCD08056661
  • Cas Number : 178261-64-8
  • Molecular Weight : 268.154

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115 - 116
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86443

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7M-599S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart