KEY087886657 Key Organics / BIONET

2-(2,5-dichlorophenyl)-2,3,4,5-tetrahydro-1,5-benzothiazepin-4-one

  • MDL number MFCD08056665
  • Cas Number : 886361-99-5
  • Molecular Weight : 324.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 233 - 235
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88291

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7Z-0210_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart