KEY087886660 Key Organics / BIONET

(2E)-3-(dimethylamino)-1-(10H-phenothiazin-2-yl)prop-2-en-1-one

  • MDL number MFCD08056668
  • Cas Number : 63285-46-1
  • Molecular Weight : 296.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245 - 247
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88304

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7Z-0819_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart