KEY087886662 Key Organics / BIONET

2-chloro-4-(3-methyl-1H-pyrazol-1-yl)benzoic acid

  • MDL number MFCD08056672
  • Cas Number : 220461-68-7
  • Molecular Weight : 236.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186 - 189
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88316

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7Z-5041_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart