KEY087886665 Key Organics / BIONET

3-amino-3-methyl-1-[3-(trifluoromethyl)phenyl]thiourea

  • MDL number MFCD08056673
  • Cas Number : 70483-65-7
  • Molecular Weight : 249.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130 - 132
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90124

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8N-661S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart