KEY087886666 Key Organics / BIONET

4-(4-methylphenyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD08056675
  • Cas Number : 886362-01-2
  • Molecular Weight : 210.236

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238 - 240
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91377

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8Z-0808_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart