KEY087886674 Key Organics / BIONET

N-[(4E)-6-chloro-3,4-dihydro-2H-1-benzothiopyran-4-ylidene]hydroxylamine

  • MDL number MFCD08056679
  • Cas Number : 1029134-50-6
  • Molecular Weight : 213.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138 - 140
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93142

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9N-378S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart