KEY087886678 Key Organics / BIONET

2,4-di-tert-butyl-6-{[(2-{[(3,5-di-tert-butyl-2-methylphenyl)methyl](methyl)amino}ethyl)(methyl)amino]methyl}phenol

  • MDL number MFCD08056682
  • Cas Number : 886362-16-9
  • Molecular Weight : 522.862

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94821

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-0008_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart