KEY087886683 Key Organics / BIONET

1-(3,4-dimethoxyphenyl)-8,9-dimethoxy-3-phenylpyrrolo[2,1-a]isoquinoline

  • MDL number MFCD08056693
  • Cas Number : 886362-22-7
  • Molecular Weight : 439.511

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94844

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-0047_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart