KEY087886686 Key Organics / BIONET

3-nitrophenyl 7-methoxy-2,2-diphenyl-2H-1,3-benzodioxole-5-carboxylate

  • MDL number MFCD08056697
  • Cas Number : 30263-87-7
  • Molecular Weight : 469.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-nitrophenyl 7-methoxy-2,2-diphenyl-1,3-benzodioxole-5-carboxylate
Inchi Code 1S/C27H19NO7/c1-32-23-15-18(26(29)33-22-14-8-13-21(17-22)28(30)31)16-24-25(23)35-27(34-24,19-9-4-2-5-10-19)20-11-6-3-7-12-20/h2-17H,1H3
Inchi Key GTHRBSBYHVVDIA-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94855

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-0061_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart