KEY087886712 Key Organics / BIONET

1-(4-chlorophenyl)-1H-1,2,3-triazole-4-carbaldehyde

  • MDL number MFCD01109282
  • Cas Number : 113934-27-3
  • Molecular Weight : 207.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151 - 153
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74280

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4F-375S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart