KEY087886725 Key Organics / BIONET

4-chloro-5-methoxy-2-(methylsulfanyl)pyrimidine

  • MDL number MFCD00223689
  • Cas Number : 87026-45-7
  • Molecular Weight : 190.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73 - 77
Purity 97
Storage temp. Store under N2 at 4°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74949

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4J-396S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart