KEY087886734 Key Organics / BIONET

3-(quinoxalin-2-yl)aniline

  • MDL number MFCD02737921
  • Cas Number : 432004-76-7
  • Molecular Weight : 221.263

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164 - 166
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76899

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0610_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart