KEY087886743 Key Organics / BIONET

5-(thiophen-3-yl)furan-2-carbaldehyde

  • MDL number MFCD06740284
  • Cas Number : 886361-73-5
  • Molecular Weight : 178.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 57 - 59
Iupac Name 5-(3-thienyl)-2-furaldehyde
Inchi Code 1S/C9H6O2S/c10-5-8-1-2-9(11-8)7-3-4-12-6-7/h1-6H
Inchi Key GYMRFFNBNAMYNM-UHFFFAOYSA-N

Description

Melting Point 57 - 59
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77502

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4Z-0704_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart