KEY087886746 Key Organics / BIONET

2-{2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid

  • MDL number MFCD08056629
  • Cas Number : 17969-63-0
  • Molecular Weight : 287.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134 - 136
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77506

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4Z-0711_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart