KEY087886750 Key Organics / BIONET

[2,3'-bithiophene]-5-carboxylic acid

  • MDL number MFCD05885496
  • Cas Number : 60141-31-3
  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 188 - 190
Iupac Name 2,3'-bithiophene-5-carboxylic acid
Inchi Code 1S/C9H6O2S2/c10-9(11)8-2-1-7(13-8)6-3-4-12-5-6/h1-5H,(H,10,11)
Inchi Key NUZWQKDXTFZYAX-UHFFFAOYSA-N

Description

Melting Point 188 - 190
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77510

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4Z-0715_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart