KEY087886752 Key Organics / BIONET

2,3,4,5,6-pentafluorophenyl 4-bromobenzene-1-sulfonate

  • MDL number MFCD00404028
  • Cas Number : 848649-38-7
  • Molecular Weight : 403.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66 - 68
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63005

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12X-0202_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart