KEY087886765 Key Organics / BIONET

3-amino-3-(5-bromo-2-fluorophenyl)propanoic acid

  • MDL number MFCD03002520
  • Cas Number : 299441-23-9
  • Molecular Weight : 262.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 218 - 220DEC
Iupac Name 3-(5-bromo-2-fluorophenyl)-beta-alanine
Inchi Code 1S/C9H9BrFNO2/c10-5-1-2-7(11)6(3-5)8(12)4-9(13)14/h1-3,8H,4,12H2,(H,13,14)
Inchi Key QHKWNVXSQJZRJG-UHFFFAOYSA-N

Description

Melting Point 218 - 220DEC
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77528

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4Z-0828_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart