KEY087886782 Key Organics / BIONET

2-(1H-pyrazol-3-yl)pyrazine

  • MDL number MFCD03618495
  • Cas Number : 111781-54-5
  • Molecular Weight : 146.153

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155 - 157
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81137

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5Z-0806_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart