KEY087886783 Key Organics / BIONET

2,3,4,5,6-pentafluorophenyl 4-methoxybenzene-1-sulfonate

  • MDL number MFCD05975127
  • Cas Number : 663175-94-8
  • Molecular Weight : 354.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58 - 60
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3490

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10X-0327_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart