KEY087886785 Key Organics / BIONET

4-[(4-methylphenyl)methoxy]benzoic acid

  • MDL number MFCD00625986
  • Cas Number : 56442-19-4
  • Molecular Weight : 242.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 216 - 218
Iupac Name 4-[(4-methylbenzyl)oxy]benzoic acid
Inchi Code 1S/C15H14O3/c1-11-2-4-12(5-3-11)10-18-14-8-6-13(7-9-14)15(16)17/h2-9H,10H2,1H3,(H,16,17)
Inchi Key GQDITHZKLNZHHU-UHFFFAOYSA-N

Description

Melting Point 216 - 218
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63086

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/12Y-0706_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart