KEY087886799 Key Organics / BIONET

4-bromo-N-(2-hydroxyethyl)benzamide

  • MDL number MFCD00456170
  • Cas Number : 57728-67-3
  • Molecular Weight : 244.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 146 - 147
Iupac Name 4-bromo-N-(2-hydroxyethyl)benzamide
Inchi Code 1S/C9H10BrNO2/c10-8-3-1-7(2-4-8)9(13)11-5-6-12/h1-4,12H,5-6H2,(H,11,13)
Inchi Key PUTNMGZYPWHWLJ-UHFFFAOYSA-N

Description

Melting Point 146 - 147
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82933

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6M-559S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart