KEY087886800 Key Organics / BIONET

2,3,4,5,6-pentafluorophenyl 3-(trifluoromethyl)benzene-1-sulfonate

  • MDL number MFCD05975128
  • Cas Number : 885950-36-7
  • Molecular Weight : 392.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point OIL
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3491

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10X-0331_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart