KEY087886809 Key Organics / BIONET

2,3,4,5,6-pentafluorophenyl 4-tert-butylbenzene-1-sulfonate

  • MDL number MFCD05975131
  • Cas Number : 885950-38-9
  • Molecular Weight : 380.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93 - 95
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3494

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10X-0341_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart