KEY087886813 Key Organics / BIONET

4-amino-1,2,5-oxadiazole-3-carboxylic acid

  • MDL number MFCD00235172
  • Cas Number : 78350-50-2
  • Molecular Weight : 129.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 220 - 222
Iupac Name 4-amino-1,2,5-oxadiazole-3-carboxylic acid
Inchi Code 1S/C3H3N3O3/c4-2-1(3(7)8)5-9-6-2/h(H2,4,6)(H,7,8)
Inchi Key YOXIXLVJYMCCIB-UHFFFAOYSA-N

Description

Melting Point 220 - 222
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84510

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6Z-0700_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart