KEY087886817 Key Organics / BIONET

4-(pyrazin-2-yl)benzaldehyde

  • MDL number MFCD02684109
  • Cas Number : 127406-08-0
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 84 - 87
Iupac Name 4-(2-pyrazinyl)benzaldehyde
Inchi Code 1S/C11H8N2O/c14-8-9-1-3-10(4-2-9)11-7-12-5-6-13-11/h1-8H
Inchi Key QEJFDPPZUXABRM-UHFFFAOYSA-N

Description

Melting Point 84 - 87
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84514

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6Z-0709_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart