KEY087886821 Key Organics / BIONET

methyl 3-amino-4-chloro-1-benzothiophene-2-carboxylate

  • MDL number MFCD01566406
  • Cas Number : 35212-86-3
  • Molecular Weight : 241.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105 - 109
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3637

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10Y-0710_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart