KEY087886830 Key Organics / BIONET

4'-chloro-3-fluoro-[1,1'-biphenyl]-4-carbonitrile

  • MDL number MFCD07366534
  • Cas Number : 885949-48-4
  • Molecular Weight : 231.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168 - 171
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3639

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10Y-0713_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart