KEY087886850 Key Organics / BIONET

5-tert-butyl-2-(chloromethyl)-1,3-benzoxazole

  • MDL number MFCD07366538
  • Molecular Weight : 223.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102 @ 0.1 mBar
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66578

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1Z-0828_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart