KEY087886853 Key Organics / BIONET

2-(chloromethyl)-6-methyl-1,3-benzoxazole

  • MDL number MFCD07366539
  • Cas Number : 143708-33-2
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 68 -70
Iupac Name 2-(chloromethyl)-6-methyl-1,3-benzoxazole
Inchi Code 1S/C9H8ClNO/c1-6-2-3-7-8(4-6)12-9(5-10)11-7/h2-4H,5H2,1H3
Inchi Key XHGXJTDZCATPBF-UHFFFAOYSA-N

Description

Melting Point 68 -70
Purity 95%
Storage temp. Store under N2
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66579

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/1Z-0830_SDS.pdf
hcodes H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart