KEY087886858 Key Organics / BIONET

4-methyl-3-(1H-pyrrol-1-yl)aniline

  • MDL number MFCD07643299
  • Cas Number : 94009-17-3
  • Molecular Weight : 172.231

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point OIL
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Brown Liquid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88309

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/7Z-0827_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart