KEY087886864 Key Organics / BIONET

(E)-N-[(4-chloro-3-nitrophenyl)methylidene]hydroxylamine

  • MDL number MFCD00160759
  • Cas Number : 66399-01-7
  • Molecular Weight : 200.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147 - 150
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67119

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2G-344S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart