KEY087886867 Key Organics / BIONET

methyl 3-benzamidothiophene-2-carboxylate

  • MDL number MFCD00665577
  • Cas Number : 79128-70-4
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99 - 101
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90052

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8N-360S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart