KEY087886872 Key Organics / BIONET

8-(benzyloxy)imidazo[1,2-a]pyridine

  • MDL number MFCD06739199
  • Cas Number : 96428-16-9
  • Molecular Weight : 224.263

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75 - 77
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91382

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8Z-0821_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart