KEY087886873 Key Organics / BIONET

N1-(2-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}ethyl)-4-(trifluoromethyl)benzene-1,2-diamine hydrochloride

  • MDL number MFCD00793061
  • Cas Number : 317822-50-7
  • Molecular Weight : 435.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67706

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2J-925_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart