KEY087886920 Key Organics / BIONET

3-chloro-N-(prop-2-en-1-yl)benzamide

  • MDL number MFCD01181345
  • Cas Number : 35306-52-6
  • Molecular Weight : 195.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44 - 47
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4924

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11J-363S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart