KEY087886930 Key Organics / BIONET

5-[(4-chlorophenyl)sulfanyl]thiophene-2-carbaldehyde

  • MDL number MFCD07698586
  • Cas Number : 139120-69-7
  • Molecular Weight : 254.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40 - 42
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70058

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2Z-0716_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart