KEY087886933 Key Organics / BIONET

2-amino-N-hydroxybenzamide

  • MDL number MFCD00456219
  • Molecular Weight : 152.153

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141 - 143
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4997

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11J-582S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart